Learn More
GUARDIAN EQUIPMENT Eyewash, Wall Mounted, AutoFlow™ 90º Swing-Down
Supplier:  GUARDIAN EQUIPMENT G1848
Encompass_Preferred
AutoFlow™ eyewash for mounting on wall. Spray heads swing down from storage to operational position, activating water flow. Available in a variety of spray head configurations to minimize obstructions at a sink. Note: If unit is not installed at a sink, floor drain should be provided underneath unit to prevent accumulation of water on floor.
Catalog No. NC2941334