Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Wall Pipette Rack Mounts for 4 Pipettes, 2 Multichannel
SDP

Catalog No. 22000777
Change view
Click to view available options
Includes:
Magnets
Screws, Anchors
VHB Tape
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-777 Screws, Anchors
22-000-776 VHB Tape
22-000-791 Magnets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-777 Supplier Poltex™ Supplier No. PIPRKWL4MUL2W
Add to Cart
Add to Cart

Pipette/Multichannel pipette holder for 4 pipettes and 2 multichannel pipettes.

  • PETG Construction
  • Fits most standard brands of pipettes
  • Choose from wall mount, powerful magnets, or VHB tape

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
Material PETG
Positions 4, 2
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipettes, Multichannel Pipettes
Length (Metric) 280 mm
Includes Screws, Anchors
Product Type Pipette Stand
Height (Metric) 104 mm
Width (Metric) 280 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.