Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Wall Pipette Rack Mounts for 5 Pipettes, 1 Multichannel
SDP

Catalog No. 22000790
Change view
Click to view available options
Includes:
Magnets
Screws, Anchors
VHB Tape
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-790 Magnets
22-000-778 VHB Tape
22-000-792 Screws, Anchors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-790 Supplier Poltex™ Supplier No. PIPRKWL5MUL1M
Add to Cart
Add to Cart

Pipette/Multichannel pipette holder that holds 5 pipettes and 1 multichannel pipette.

  • Made of durable PETG Construction
  • Fits most standard brands of pipettes
  • Choose from wall mount (screws and anchors included), powerful magnets, or VHB tape

Specifications

Color Clear
Holds Pipettes
Material PETG
Positions 5, 1
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipettes, Multichannel Pipettes
Length (Metric) 280 mm
Includes Magnets
Product Type Pipette Stand
Height (Metric) 104 mm
Width (Metric) 280 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.