Learn More
Walter Products™ Enlarged Liver Model
SCI_ED
Catalog No. S07143 Shop All Walter Products Products
Change view
Click to view available options
Dissects into three parts
Catalog No.
S07143 Dissects into three parts
1 options
Catalog No. S07143 Supplier Walter Products™ Supplier No. B10408
Displays left, right, caudate and quadrate lobes of human liver.
  • Enlarged approximately five times
  • Shows gall bladder, extra hepatic gall ducts and bile ducts system
  • Made of durable plastic and dissects into three parts
  • Mounts on plastic base
Specifications
For Use With (Application) Displays left, right, caudate and quadrate lobes of Human liver, enlarged approximately five times and shows the left, right, caudate and quadrate lobes of the Human liver
Includes Gall bladder, extra hepatic gall ducts and bile ducts system
Material Plastic
Size Enlarged
Product Type Model, Liver