Learn More
Walter Products™ Half Size Human Skeletons
SCI_ED
Catalog No. S07106 Shop All Walter Products Products
Change view
Click to view available options
Product Type:
Model, Human Skeleton w/ Muscles
Model, Human Skeleton w/ Nerve Ending
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07106 Model, Human Skeleton w/ Muscles
S07105 Model, Human Skeleton w/ Nerve Ending
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S07106 Supplier Walter Products™ Supplier No. B10204
Add to Cart
Add to Cart
Includes
Labeled diagram
Wired teaching skeleton for natural movement.
  • Three-foot plastic skeleton resting on heavy metal stand
  • Removable skull with calvarium cut
  • Mandible hinged with springs in place to examine TMJ problems
  • Arms and legs held on by small screws, making it easy to bend extremities in all directions
  • All bones casted individually, except in hands and feet
Specifications
For Use With (Application) Teacher tool
Includes Muscle origins in red and muscle insertions in blue, movable legs, arms and skull, colored diagram
Material Plastic
Mounting Stand
Size Half-Size
Product Type Model, Human Skeleton w/ Muscles