Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Life-Size Digestive System Model
SCI_ED

Catalog No. S07123 Shop All Walter Products Products
Change view
Click to view available options
Dimension: 35.4 x 12 x 5.1 in. (90 x 30 x 13cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07123 Dimension: 35.4 x 12 x 5.1 in. (90 x 30 x 13cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07123 Supplier Walter Products™ Supplier No. B10420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Illustrates human digestive system from mouth cavity to rectum.

  • Shows oral cavity, pharynx, esophagus, liver with gall bladder, and pancreas
  • Open stomach along frontal plane
  • Duodenum, caecum, and rectum of digestive system are open to expose interior
  • Removable transverse colon for further examination
  • Dimensions: 35.4 x 12 x 5.1 in. (90 x 30 x 13cm)

Specifications

For Use With (Application) Illustrates Human digestive system from mouth cavity to rectum
Includes Oral cavity, pharynx, esophagus, liver with gall bladder and the pancreas
Product Type Model, Digestive System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.