Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products M Series Microscopes
SCI_ED

Catalog No. S193201X Shop All Walter Products Products
Change view
Click to view available options
Objective Types:
DIN
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Objective Types
S193201X DIN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S193201X Supplier Walter Products Supplier No. MCXM100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unique base design provides comfort while viewing and stability while operating this instrument.

  • Calibrated pointer provides actual measurements and a reverse nosepiece on the stage makes the M Series more comfortable to use and provides more working room in the stage area
  • Suitable for grades 11 through College and University level applications as well as a great instrument for industry and lab use
  • DIN objectives 4X, 10X, 40XR and 100XR (R: Retractable) are parfocaled and parcentered
  • Models are available in tungsten illumination
  • Choice of monocular, 90° video, or binocular head
  • Heads rotate 360° for convenient viewing
  • Quadruple reverse nosepiece

Warranty and Services

Limited lifetime warranty

Specifications

Axis Type Coaxial
Condenser Type 1.25 N.A. Abbe
Design Type Mechanical
Diaphragm Iris
Eyepiece WF10X
Head Style Monocular
Lighting Type LED
Magnification Power 4X, 10X, 40XR, 100XR (oil)
Objective Types DIN
For Use With (Application) Industry and lab use
Grade University
Includes Vinyl dustcover and instruction manual, calibrated pointer, quadruple reverse nosepiece
Nosepiece 4-position reversed
Warranty Limited lifetime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.