Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WAPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538024
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538024 Supplier Invitrogen™ Supplier No. PA538024
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is A-20 cell lysate. At least four isoforms of WAPL are known to exist.

Transcriptional repressor which may play a role in development of the central nervous system (CNS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen WAPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene WAPL
Gene Accession No. Q65Z40, Q7Z5K2
Gene Alias A530089A20Rik; BC037674; DIF-2; Dioxin-inducible factor 2; FOE; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); friend of EBNA2 protein; KIAA0261; RGD1308960; WAPAL; WAPL; WAPL cohesin release factor; wings apart-like homolog (Drosophila); wings apart-like protein homolog
Gene Symbols WAPL
Host Species Rabbit
Immunogen 18 amino acid peptide near the amino terminus of human WAPL.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218914, 23063, 290577
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.