Learn More
Lkt Laboratories CAS 81-81-2, ≥98%, VKORC1 inhibitor used to prevent blood clot formation and migration. It inhibits the formation of coagulation factors II, VII, IX, and X.
SDP
Supplier:  Lkt Laboratories W0269
Encompass_Preferred
Warfarin is an orally available antithrombotic commonly used for preventing blood clot formation and migration. The S enantiomer of warfarin is somewhat more potent than the R enantiomer. This coumarin inhibits the subunit 1 of the vitamin K epoxide reductase complex (VKORC1), depleting the reduced form of vitamin K (KH2) and inhibiting the formation of coagulation factors II, VII, IX, and X. Warfarin has a relatively slow onset and requires 1 to 2 days to take effect. This compound is highly sensitive to a wide variety of genetic polymorphisms and is metabolized by cytochrome P4502C9; it also interacts with many other medications and foods, requiring diligent activity monitoring.
Catalog No. NC2535227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.