Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Warfarin, MilliporeSigma™ Supelco™

Catalog No. 111017779 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7779 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7779 Supplier MilliporeSigma Supelco Supplier No. A225010G
Environmental and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 81-81-2
Physical Form Neat
Linear Formula C19H16O4
MDL Number MFCD00006854
Melting Point 162°C to 164°C
Synonym 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin; Coumafene
Type Analytical Standard
Quantity 10 g
Formula Weight 308.33
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.