Learn More
Cytiva WASH VALVE FOR AKTA START
Supplier:  Cytiva 29023897
Encompass
Wash Valve for KTA Start For use with the AKTA start automated protein purification system. The wash valve is used to divert the flow path to the waste when needed.
Catalog No. NC1442654