Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Waste Disposal Labels - Coated Paper

Catalog No. 19110991 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
EMPTY
THIS CONTAINER HAS BEEN TRIPLE RINSED
USED BATTERIES Hazardous Waste
USED OIL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-110-991 EMPTY
19-107-312 THIS CONTAINER HAS BEEN TRIPLE RINSED
19-110-992 USED BATTERIES Hazardous Waste
19-110-994 USED OIL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-110-991 Supplier Brady™ Supplier No. 60363
Add to Cart
Add to Cart

Use these durable labels to easily identify and sort waste for disposal.

  • All HazMat Shipping Labels meet D.O.T. Requirements HM-181 Regulation.
  • B-235 Coated Paper with pressure sensitive acrylic adhesive from Brady™ adheres to most surfaces
  • Labels remove cleanly, leaving no residue on surface
  • Operating temperature of -40°F to 150°F (-40°C to 65.5°C)

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Color Black on White
Finish Semi-gloss
Format Label
Material Paper
Printable Sides Single-side Printable
Product Type Label
Display Text EMPTY
Languages English
Quantity 100/Pk.
Shape Rectangle
Size (English) 6 x 6 in.
Dimensions (L x W) 6 x 6 in.
Width (English) 6 in.
Width (Metric) 158.75 mm
Height (English) 6 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.