Learn More
MCR Safety Hi-Vis Orange Nylon Work Gloves with Foam Nitrile Coated Palm
Catalog No. 19812708 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-708 Small
19-812-707 Medium
19-812-706 Large
19-812-709 X-Large
19-812-710 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-708 Supplier MCR Safety Supplier No. 96790HVIS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
A printed safety reminder on the back of each glove reminds users to watch their hands at all times. Hi-vis orange nitrile foam on the palm provides abrasion resistance and enhances wet/dry grip capabilities.
  • Nylon shell is comfortable and flexible for optimal dexterity
  • Hi-vis orange BNF (Breathable Nitrile Foam) with NFT™ coating on the palm and fingertips
  • Nitrile foam provides abrasion resistance and enhanced grip capabilities
  • Watch Your Hands logo printed on the back of each glove as a constant safety reminder
  • ActiFresh™ treatment reduces bacteria and odors
Specifications
ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile Foam
Color Hi-vis Orange, Gray
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
Finger Style Full
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 96790HVI
Yarn Gauge 15
Show More Show Less
WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product