Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Solstice

Catalog No. 6003081 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Glass Bottle, Gl45
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600-30-81 4 x 4 L Glass Bottle
600-30-79 6 x 1 L Glass Bottle, Gl45
600-30-80 4 x 2.5 L Glass Bottle, Gl45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 600-30-81 Supplier Solstice Supplier No. LC4524
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 64-18-6 , 7732-18-5
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

CAS Max % 99.9000%, 0.1000%
Density 0.997 g/cm3
Quantity 4 x 4 L
Packaging Glass Bottle
Formula Weight 46.03
Grade HPLC
Chemical Name or Material Water with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC
ShelfLife 365 days from date of manufacture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.