Learn More
Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Solstice
Shop All Honeywell Research Chemicals ProductsSpecifications
Specifications
| Boiling Point | 100°C |
| CAS Max % | 99.9000%, 0.1000% |
| Density | 0.9982 g/cm3 |
| Grade | HPLC |
| Packaging | Glass Bottle, Gl45 |
| Quantity | 4 x 2.5 L |
| Chemical Name or Material | Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.