Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water 50%, Acetonitrile 50%, LabReady™ Solvent Blend, for LC-MS and HPLC, Solstice

Catalog No. 6000164 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-001-64 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-001-64 Supplier Solstice Supplier No. LC4354
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 75-05-8 , 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name ethanol
SMILES O

Specifications

CAS Min % 50.0000%, 43.9000%
CAS Max % 56.1000%, 50.0000%
Density 0.7857 g/cm3
Quantity 4 x 4 L
Flash Point 3°C
Packaging Glass Bottle
UN Number UN1648
Boiling Point 82°C
Grade HPLC
Chemical Name or Material Water 50%, Acetonitrile 50%, LabReady™ Solvent Blend, for LC-MS and HPLC
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.