Learn More
New Pig Water-Absorbing Sock
SDP
Catalog No. 18389160 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Cellulose
Catalog No. Material
18-389-160 Cellulose
1 options
This item is not returnable. View return policy
Catalog No. 18-389-160 Supplier New Pig Supplier No. WTR020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Super absorbent sock absorbs and locks in large volumes of water-based leaks and spills quickly.
  • Pulls in large volumes of water on contact to confine/contain pools and puddles and prevent damage and slip and fall injuries; fast-wicking sock saves time on cleanup
  • Super absorbent polymer (SAP) filler and spunbound sock skin lock water and water-based liquids inside without leakage
  • Sock retains and absorbs 15 times its weight in liquid so it can act as a weighted berm when saturated
  • Great for indoor applications where water needs to be quickly absorbed or diverted; soaks up water from windows, cracked foundations and seepage under doors
  • Great for surrounding machines during system breakdowns; use around pipes, broken hoses, leaking hot water tanks or malfunctioning equipment
  • Soaks up excess washdown water in food service and food processing areas
Specifications
Material Cellulose