Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Grade, Vion™

Catalog No. 11178112 Shop All Aldon Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-178-112 500 mL
11-178-109 125 mL
11-178-110 1 L
11-178-111 250 mL
11-178-113 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-178-112 Supplier Aldon Corporation Supplier No. VNWW7000500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vion ACS Reagent grade water is guaranteed to meet the purity standards set by the American Chemical Society (ACS). It is a high-quality, highly purified water with minimal contaminants, and is ideal for use in laboratory settings for sensitive chemical analyses where extreme purity is required.

Specifications:

  • Conductivity ≤ 2.0 μS/cm
  • Silicate (as SiO2) ≤ 0.01 ppm
  • Heavy Metals (as Pb) ≤ 0.01 ppm
  • Substances Reducing Permanganate - Passes Test
  • Chloride (Cl) ≤ 0.4 ppm
  • Nitrate (NO3) ≤ 0.4 ppm
  • Phosphate (PO4) ≤ 1 ppm
  • Sulfate (SO4) ≤ 1 ppm
  • UV A300 nm ≤ 0.005
  • UV A254 nm ≤ 0.005
  • UV A210 nm ≤ 0.010
  • UV A200 nm ≤ 0.010
  • LC Absorbance - Passes Test
  • LC Gradient Elution - Passes Test

Chemical Identifiers

Molecular Weight (g/mol) 18.02 g/mol

Specifications

CAS 7732-18-5
Percent Purity 100%
Grade ACS
Quantity 500 mL
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.