Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Reagent, Solstice

Catalog No. 60046041 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-041 4 L Poly Bottle
60-046-040 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-041 Supplier Solstice Supplier No. 3200724L

missing translation for 'validMainframeMsg'

ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade ACS Reagent
Packaging Poly Bottle
Density 1.000g/mL at 3.98°C
Quantity 4 L
Chemical Name or Material Water
EINECSNumber 231-791-2
RTECSNumber ZC0110000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.