Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ASTM Type I, Vion™

Catalog No. 11178118 Shop All Aldon Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-178-118 60 mL
11-178-114 125 mL
11-178-115 1 L
11-178-116 250 mL
11-178-117 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-178-118 Supplier Aldon Corporation Supplier No. VNWW700160ML
Only null left
Add to Cart
Add to Cart

Vion ASTM Type I water meets the strictest standards defined by the American Society for Testing and Materials (ASTM). It is ideal for a wide range of ultra-sensitive laboratory uses including gas chromatography, HPLC, mass spectrometry, atomic absorption spectroscopy, and trace analysis.

Specifications:

  • Resistivity ≥ 18 MOhm
  • Conductivity ≤ 0.056 μS/cm
  • Total Organic Carbon (TOC) ≤ 50 ppb
  • Silicate (as SiO2) ≤ 3 ppb
  • Chloride (Cl) ≤ 1 ppb
  • Sodium (Na) ≤ 1 ppb
  • Endotoxin ≤ 0.03 EU/mL
  • Heterotrophic Bacteria Count (HBC) ≤ 10 CFU/L

Chemical Identifiers

Molecular Weight (g/mol) 18.02 g/mol

Specifications

CAS 7732-18-5
Percent Purity 100%
Quantity 60 mL
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.