Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ASTM Type II, Reagents

Catalog No. 01336891
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-891 4 L
01-336-895 10 L
01-336-894 1 L
01-336-892 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-336-891 Supplier Reagents Holdings Llc Supplier No. C23462004A
Add to Cart
Add to Cart

Water, ASTM Type II. ACS Reagent Meets ASTM requirements for Distilled Water, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Packaging Poly Bottle
Sterility Non-sterile
Quantity 4 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, ASTM Type II / ACS Reagent Meets ASTM requirements for Distilled Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.