Learn More
Chemglass Life Sciences 7L Water Bath, Shallow, General Purpose, Digital, 120V
SDP
Supplier:  Chemglass Life Sciences CLS4950006
Encompass_Preferred
7L Water Bath, Shallow, General Purpose, Digital, 120V. The CLS-4950 Series high-performance water baths are accurate, easy to use, safe and durable. The digital set / digital read P.I.D. temperature controller maximizes rapid heat-up without temperature overshoot. A microprocessor achieves precise temperature control regardless of how the unit is loaded. It has a unique lid design that allows condensation to drain back into the tank without spilling. There is a gasket around the water bath tank to prevent water from dripping onto the controls. A warm air jacketed design radiates heat to the sides & bottom of the tank, eliminating hot spots. The unit has a non-contact, recessed heating element that will prevent burnouts. The water bath tanks are constructed of stainless steel and do not have seams that could leak. The unit's housings have a high quality textured powder coat finish that eliminates rust & chipping and is resistant to nicks, scratches & corrosives.
Catalog No. 50-143-474
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.