Learn More
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Absorbance | max. 0.005 at 400 nm, max. 0.005 at 250 nm, max. 0.01 at 210 nm, max. 0.01 at 205 nm |
| Boiling Point | 100°C |
| CAS Max % | 100.0000% |
| Linear Formula | H2O |
| Beilstein | 2050024 |
| Formula Weight | 18.02 |
| Grade | HPLC |
| Packaging | Glass Bottle |
| Density | 1.000 g/mL at 3.98 °C(lit.) |
| Quantity | 4 x 4 L |
| Show More |
Safety and Handling
| ShelfLife | 720 days from date of manufacture |
| EINECSNumber | 231-791-2 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.