Learn More
Supertek™ Water Clock Kit
GREENER_CHOICESCI_ED
Turn water bottles into a battery to power the digital clock with this Water Clock Kit
Supplier:  Supertek™ PH96024S3
Catalog No. S01267
Only null left
Add to Cart
Description
Description
  • Explore what else can power the digital clock!
  • Comes with zinc and copper electrodes
  • 1 Digital clock
  • Instructions are included
  • Ages 7 plus
Specifications
Specifications
Potato Clock
Digital
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.