Learn More
Gibco™ Water For Injection (WFI) for Cell Culture

Description
- Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
- The water is prepared by distillation and filtration (0.1 μm), with no added substances
- In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
- Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
- It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Should be stored at 15° to 30°C
Specifications
Specifications
| pH | 4 to 6 |
| Grade | Cell Culture |
| Sterility | Sterile-filtered |
| Treatment(s) | None |
| Quantity | 10 x 500 mL |
| Purification Method | Distilled and filtered (0.1 μm) |
| Shipping Condition | Room Temperature |
| Product Line | Gibco |
Frequently Asked Questions (FAQs)
These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.
Water for Injection (WFI) for Cell Culture is prepared by distillation and filtration (0.1 µm), with no added substances. It is labeled For Research Use Only of Futher Manufacturing. Not for use in diagnostic procedures.
Safety and Handling
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.