Learn More
Water Hardness Indicator, 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| pH Range | Alkaline |
| Volume (Metric) | 500 mL |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Test Method | APHA/EPA for Water Hardness |
| Type | Water Hardness Indicator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.