Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water for Injection, USP, Sterile Grade, Cytiva Hyclone™

Catalog No. WFIUSP10L Shop All Cytiva Products
Change view
Click to view available options
Quantity:
6 x 500 mL
10 L
6 x 1 L
1 x 20 L
200 L
1 x 100 L
Packaging:
Bioprocess Bag
PET Plastic Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
WFI-USP-10L 10 L Bioprocess Bag
WFI-USP-1X6 6 x 1 L PET Plastic Bottle
WFI-USP-20L 1 x 20 L Bioprocess Bag
WFI-USP-2XL 200 L Bioprocess Bag
WFI-USP-5XM 6 x 500 mL PET Plastic Bottle
WFI-USP-1XL 1 x 100 L Bioprocess Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. WFI-USP-10L Supplier Cytiva Supplier No. WFIUSP10L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Water for Injection, USP, is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer.

  • Complies with the USP monograph for water for injection packaged in bulk for commercial use
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in applications where a high quality sterile water is required

For Manufacturing or Research Use only, not intended for Diagnostic or Therapeutic Use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade USP
Packaging Bioprocess Bag
Sterility Sterile
Certificate of Analysis Yes
Quantity 10 L
ShelfLife 36 Months
Recommended Storage 2-30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.