Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water for Irrigation, USP, Sterile Grade, Cytiva Hyclone™

Catalog No. WIRUSP1X6 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Bioprocess Bag
HDPE Jug
PET Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
WIR-USP-1X6 6 x 1 L PET Plastic Bottle
WIR-USP-5XM 6 x 500 mL PET Plastic Bottle
Z0304 4 x 4 L HDPE Jug
WIR-USP-20L 20 L Bioprocess Bag
WIR-USP-2XL 200 L Bioprocess Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. WIR-USP-1X6 Supplier Cytiva Supplier No. WIRUSP1X6
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Water for Irrigation, USP is a sterile, hypotonic, non-pyrogenic fluid or solvent. It is prepared by terminal distillation and contains no antimicrobial or bacteriostatic agents or added buffers.

  • The pH is 5.0 to 7.0
  • Complies with the USP monograph for water for irrigation
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in applications where a high quality sterile water is required

For Manufacturing or Research Use only, not intended for Diagnostic or Therapeutic Use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade USP
Packaging PET Plastic Bottle
Sterility Sterile
Certificate of Analysis Yes
Quantity 6 x 1 L
ShelfLife 36 Months
Recommended Storage 2-30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.