Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, CHROMASOLV™ LC-MS, Solstice

Catalog No. 60090634 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-634 4 x 2.5 L
60-046-746 4 L
60-046-747 500 mL
60-046-744 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-090-634 Supplier Solstice Supplier No. 392534X2.5L

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Absorbance max. 0.022 at 210 nm, max. 0.004 at 230 nm
Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor
IUPAC Name water
Molecular Weight (g/mol) 18.02
Formula Weight 18
Grade LC-MS
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 4 x 2.5 L
Chemical Name or Material Water, CHROMASOLV™ LC-MS
Show More Show Less
ShelfLife 547 days from date of manufacture
EINECSNumber 231-791-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.