Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, LC-MS Grade, For Chromatography LiChrosolv™, MilliporeSigma™

Catalog No. 01100249 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-249 2.5 L
01-100-248 1 L
01-100-250 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-100-249 Supplier MilliporeSigma Supplier No. 1.15333.2500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Distilled water, Deionized water
IUPAC Name water
SMILES O

Specifications

pH 6 to 8 (at 25°C)
Boiling Point 100°C (1013 hPa)
Packaging Glass Bottle
Vapor Pressure 23 hPa at 20 °C
Density 1 g/cm3
Quantity 2.5 L
Recommended Storage Store at 2 to 30 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.