Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Water Model Kit
SCI_ED

Catalog No. S41137 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S41137 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S41137 Supplier Eisco™ Supplier No. CH0603D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water Model Kit provides a tangible and hands-on display of the molecular properties of water.

  • Includes (62) white hydrogen, (31) red oxygen, (44) grey links, and (62) white links plus storage case
  • Assembly required.
  • Elements identified by color
  • For teacher use or student use

Warranty and Services

1 year limited manufacturer warranty

Specifications

Color Red, Gray, White
Height (Metric) 4.45 cm
Includes 199 pieces: 62 white Hydrogen balls, 31 red Oxygen balls, 44 gray links, 62 white links
Length (Metric) 18.4 cm
Material Plastic
Quantity 1 Set
Weight (Metric) 0.36
Width (Metric) 14 cm
Product Type Molecular Model Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.