Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Water Purification Systems Storage Reservoirs

Catalog No. 10451109
Change view
Click to view available options
Type:
Feed Water Tank
Sterile Overflow
Storage Reservoir
Capacity (Metric):
100 L
30 L
30 to 100 L
60 L
Includes:
Level Display
Level Display, Recirculation Pump
N/A
Reservoir
Tank
For Use With (Equipment):
GenPure, GenPure Pro, GenPure xCAD
Pacific RO, Pacific TII
Pacific TII
Smart2Pure
Smart2Pure 12
Smart2Pure, Pacific and LabTower Water Purification Systems
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity (Metric) Includes For Use With (Equipment)
10-451-109 Sterile Overflow 30 to 100 L N/A Smart2Pure, Pacific and LabTower Water Purification Systems
10-451-007 Storage Reservoir 30 L Level Display Pacific RO, Pacific TII
10-451-190 Sterile Overflow 30 to 100 L N/A Smart2Pure
13-998-287 Storage Reservoir 60 L Level Display, Recirculation Pump Pacific TII
13-998-286 Storage Reservoir 30 L Level Display, Recirculation Pump Pacific TII
10-451-015 Storage Reservoir 60 L Reservoir Smart2Pure 12
10-451-010 Storage Reservoir 30 L Reservoir Smart2Pure 12
10-451-009 Feed Water Tank 30 L Tank GenPure, GenPure Pro, GenPure xCAD
13-998-289 Storage Reservoir 100 L Level Display Pacific RO, Pacific TII
13-998-288 Storage Reservoir 100 L Level Display, Recirculation Pump Pacific TII
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10-451-109 Supplier Thermo Scientific™ Supplier No. 50132714
Add to Cart
Add to Cart

Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.

Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.

Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 to 100 L
For Use With (Equipment) Smart2Pure, Pacific and LabTower Water Purification Systems
Type Sterile Overflow
Includes N/A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.