Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap White SMS Lab Coats, Open Wrists

Catalog No. 19168280E Shop All Cellucap Products
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-280E 2X-Large
19-168-280D X-Large
19-168-280F 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-168-280E Supplier Cellucap Supplier No. 35032X
Add to Cart
Add to Cart
This item is not returnable. View return policy

SMS lab coats feature open wrists and flat collar. Ideal in heavy dust or splash environments.

  • Soft and comfortable lab coat
  • SMS is non-woven, two layers of spunbonded polypropylene enhanced with an inner layer
  • Ideal for dusty areas
  • Splash protection
  • No pockets, open wrists, flat collar, white

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Dust, Splash
Style Standard Length
No. of Pockets None
Collar Type Flat
Model 3503
For Use With (Application) For dusty areas and splash protection
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.