Learn More
Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Formula Weight | 18.02 |
| Grade | Molecular Biology |
| Packaging | Nalgene Bottle |
| Density | 1 |
| Quantity | 25 mL |
| Specific Gravity | 1.000 |
| Physical Form | Liquid |
| Chemical Name or Material | Water |
Safety and Handling
| EINECSNumber | 231-791-2 |
| RTECSNumber | ZC0110000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.