Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure

Catalog No. AAJ70783AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
10 x 1 mL
500 mL
1 L
Packaging:
Nalgene Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ70783AE 100 mL Nalgene Bottle
AAJ70783XCR 10 x 1 mL Nalgene Bottle
AAJ70783AC 25 mL Nalgene Bottle
AAJ70783AP 500 mL Nalgene Bottle
AAJ70783K2 1 L Nalgene Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAJ70783AE Supplier Thermo Scientific Supplier No. J70783AE
Add to Cart

CAS: 7732-18-5 | H2O | 18.015 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Bottle
Density 1
Quantity 100 mL
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water
EINECSNumber 231-791-2
RTECSNumber ZC0110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.