Learn More
Water Rocket and Class Packs

Description
Requires
Standard bicycle pump with a pressure gauge; assembly of foam fins requires low temperature hot glue gun
Packs consist of:
- Pre-cut rigid foam fin sets
- Translucent molded nozzles
- Foam recovery bumpers
- Color assembly and use instructions
Specifications
Specifications
| Product Type | Water Rocket Class Pack |
| Includes | 25 Precut rigid Foam fin set, 25 translucent molded nozzles, 25 Foam recovery bumpers, color assembly and use instructions |
| For Use With (Application) | Perfect low cost flying rocket without the hassles of traditional solid propellant rockets! Theres no fire danger (the rocket fuel is water) and no HazMat fees to ship them! |
| Subject | Physics |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.