Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Water Sampler
SDPSCI_ED

Catalog No. S89538 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Capture Water Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S89538 Capture Water Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89538 Supplier United Scientific™ Supplier No. UNWTRSMPL01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Capture water samples from specific depths with the United Scientific™ Water Sampler.

  • Capture water samples from specific depths with the United Scientific Water Sampler. A unique design allows for hassle-free sample collection: when the desired depth is reached, students simply jerk the attached line to close the apparatus.
    • Includes PVC collection tube, rubber seals and quick-release pin, Styrofoam winding float with 20 m line
    • Made from impact-resistant PVC, with soft rubber seals that hold up to 1.5 liters of liquid
    • Unit is 30 cm long, 10 cm in diameter and weighs 2 pounds
    • Activity guide included

Specifications

For Use With (Application) Capture Water Samples
Product Type Water Sampler
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.