Learn More
Grainger Water Tank Fiberglass 40.0 gal 40 psi
Supplier:  Grainger 16X847
Encompass_Preferred
The Dayton 16X847 is a pre-charged water tank with a capacity of 2 gallons. It has a maximum pressure of 125 PSI and is made from welded steel with a durable powder coat finish. This air tank features a 1/4" NPT inlet and outlet and can be used for a variety of applications including storage, transfer, and dispensing of air or water. With dimensions of 10" diameter x 17" tall and a shipping weight of 7 lbs, this compact tank provides reliable compressed air or water storage.
Catalog No. 50-276-7830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.