Learn More
Grainger Water Tank Fiberglass 80.0 gal 40 psi
Supplier:  Grainger 16X849
Encompass_Preferred
The Grainger website features fiberglass well water tanks and fittings. These tanks are designed for residential and light commercial applications and are made of fiberglass reinforced polyester for corrosion resistance. They feature stainless steel fittings and are available in capacities ranging from 30 to 120 gallons. The tanks operate with jet, submersible, and shallow well pumps to provide efficient water storage and delivery for homes and businesses. With proper installation and maintenance, these fiberglass tanks offer a durable and reliable option for well water systems.
Catalog No. 50-276-6843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.