Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with0.1% Formic Acid, LC-MS Ultra, Solstice CHROMASOLV™

Catalog No. 60044849 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-849 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-849 Supplier Solstice Supplier No. 142812L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ LC-MS Ultra, LabReady™ Solvent Blend, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
MDL Number MFCD00011332

Specifications

Quantity 2 L
Packaging Glass Bottle
UN Number NONH for all modes of transport
Formula Weight 18.02g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Water With 0.1% Formic Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.