Description
- Bi-level surface effectively scrapes dirt and moisture from shoes, trapping it beneath shoe level to prevent tracking
- Reinforced rubber nubs prevent pile from crushing and extend the performance life of the mat
- Solution-dyed polypropylene fabric is resistant to staining, dries quickly, and will not fade or rot
- Certified high-traction by the National Floor Safety Institute (NFSI)
- Available in 18 colors with choice of 4 patterns: waffle, diamond, diagonal, or geometric
- Available with choice of 100-mil or 200-mil SBR rubber
Specifications
Specifications
| Length (English) | 18 in. |
| Style | Square pattern |
| Certifications/Compliance | NFSI Certified. Made in USA |
| Thickness (English) | 7/16 in. |
| Quantity | 1 Ea. |
| Type | Entrance Mat |
| Color | Medium Gray |