Learn More
Oakton™ Waterproof pHTestr™ BNC
SCI_ED
Catalog No. S68161 Shop All Oakton Products
Change view
Click to view available options
pHTestr BNC Waterproof
Catalog No.
S68161 pHTestr BNC Waterproof
1 options
Catalog No. S68161 Supplier Oakton Supplier No. WD3562414/EMD
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Includes
Meter comes with batteries and instructions
Uses any pH electrode with BNC connector
Connects to any pH electrode with a BNC connector. Unit can be customized for wide range of applications based on selected electrode.
  • Waterproof, dustproof, floatable housing is certifiable to IP67 standards
  • Digital readout and push-button calibration for up to three pH points (4.01, 7.00, 10.00) or NIST buffers
  • Functions include HOLD, auto shutoff, and error messages
Note: Electrode not included, order separately.
Order Info
Dimensions: 6 1/2L x 1 1/2 in. dia. (16.5 x 3.8cm). Electrical requirements: four 1.5V alkaline batteries (Type A76) or silver oxide batteries (Type 303).
Certifications
CE marked CE listed.
Warranty and Services
One-year warranty