Learn More
MilliporeSigma™ Supelco™ Waters™ Fittings size 1/16 in.
Catalog No. 111004418 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
11-100-4418 5/Pk.
1 options
Catalog No. 11-100-4418 Supplier MilliporeSigma™ Supelco™ Supplier No. 58458
Add to Cart
Add to Cart
Waters™ Fittings are used for connecting most HPLC columns to a 1/16 in. tubing system.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 5/Pk.
Description 1/16 in.
Waters is a registered trademark of Waters