Learn More
Waters Corp Spherisorb ODS1 Cartridge, 80Å, 5 µm, 4 mm X 250 mm, 1/pk
Supplier:  Waters Corp PSS845552
Encompass_Preferred
Spherisorb ODS1 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS1 packing features low ligand density with a specified carbon load of 6.2%.
Catalog No. 50-784-708
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.