Learn More
MilliporeSigma™ Supelco™ Waters™ Union size 1/16 in., Stainless Steel
Catalog No. 111004639 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-4639 1/Pk.
1 options
Catalog No. 11-100-4639 Supplier MilliporeSigma™ Supelco™ Supplier No. 58289
Add to Cart
Add to Cart
Waters™ Union is used for connecting most HPLC columns to a 1/16 in. tubing system.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description 1/16 in.
Material Stainless Steel
Waters is a registered trademark of Waters