Learn More
Eisco™ Wave Apparatus
SCI_ED
Catalog No. S12967 Shop All Eisco Products
Catalog No. S12967 Supplier Eisco™ Supplier No. PH0712N8
Add to Cart
Add to Cart
Demonstrate both longitudinal and transverse wave motions. The EISCO Wave Apparatus consists of a number of eccentric discs supporting a series of metal rods revolving on a handle. Transverse waves are obtained from the straight rods and longitudinal waves are obtained from the bent rods.
  • Demonstrate longitudinal and transverse waves
Specifications
Product Type Wave Apparatus
For Use With (Application) Demonstrating both longitudinal and transverse wave motions