Learn More
Eisco™ Wave Demonstration Apparatus
SCI_ED
Catalog No. S23751 Shop All Eisco Products
Change view
Click to view available options
L x W x H: 16.9 x 5.9 x 8.7 in.
Catalog No.
S23751 L x W x H: 16.9 x 5.9 x 8.7 in.
1 options
Catalog No. S23751 Supplier Eisco™ Supplier No. PH0712
Add to Cart
Add to Cart
Demonstrates both longitudinal and transverse wave motions.
  • Eccentric discs support metal rods revolving on handle to obtain transverse waves
  • Longitudinal waves obtained from bent rods
Specifications
Product Type Wave Demonstration Apparatus
For Use With (Application) Demonstrates both longitudinal and transverse wave motions
Dimensions (L x W x H) 16.9 x 5.9 x 8.7 in.