Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Wave Demonstrator Set
SDPSCI_ED

Catalog No. S06685 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06685 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06685 Supplier United Scientific™ Supplier No. WDSET1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Wave Demonstrator Set includes two helical springs and a length of cotton string.

  • Long, narrow spring measures 0.75 x 72 in., and can be extended to 5 m.
  • Larger spring measures 3 x 4 in. and can be extended many times its length without distortion.

Order Info

Weight: 2.5 lb. (1.1kg).

Specifications

Grade 5+
Format Set
Product Type Wave Demonstrator Set
Includes 2 Helical Springs, Length of Cotton String, Long Narrow Spring, Larger Spring, Teacher Activity Guide
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.