Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wax Paraffin, Pastilles, Spectrum™ Chemical
SDP

Catalog No. 18607736 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 kg
12 x 1 lb.
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-736 12 x 1 lb. Poly Bottle
18-607-737 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-736 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W10611LBCS12
Add to Cart
Add to Cart

W1061, 8002-74-2

Spectrum™ Chemical Wax Paraffin, Pastilles consists of a mix of hydrocarbon molecules and is derived from coal, petroleum, or oil shale. It can be used in electrical insulation, lubrication and candles. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8002-74-2
Molecular Formula C21H27NO3
Molecular Weight (g/mol) 341.45
InChI Key JWHAUXFOSRPERK-UHFFFAOYNA-N
IUPAC Name 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
SMILES CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 12 x 1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.