Learn More
MARSHALL SCIENTIFIC Polyscience WBE20A11B 20L Digital Water Bath
SDP
Supplier:  MARSHALL SCIENTIFIC WBE20A11B
Encompass_Preferred
The Polyscience WBE20A11B 20L Digital Water Bath is designed to handle a wide range of laboratory procedures including incubation, inactivation and agglutination as well as most pharmaceutical, serological, biomedical and industrial procedures. To ensure optimum temperature uniformity, your Water Bath features an energy efficient, large-area heater and thermostatic control. Its PID microprocessor control (Proportional Integral Derivative) system provides proportional heat control by anticipating the approach to your set point temperature and preventing overshoot. A redundant safety thermostat is standard on all models.
Catalog No. NC2591251
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.